accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHOHOOHOHOH

CAS RN 4877-80-9

IUPAC Name: triphenylene-2,3,6,7,10,11-hexol
Synonyms: triphenylene-2,3,6,7,10,11-hexol
Molecular Weight (g/mol): 324.29
Molecular Formula: C18H12O6
InChi Key: QMLILIIMKSKLES-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C2C(=C1)C1=CC(O)=C(O)C=C1C1=CC(O)=C(O)C=C21
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)