Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6,7,10,11-Hexahydroxytriphenylene, Spectrum™ Chemical
SDP

Catalog No. 18620851 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-851 50 g
18-620-849 1 g
18-620-850 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-851 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H178650GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: Triphenylene-2,3,6,7,10,11-Hexaol, Triphenylene-2,3,6,7,10,11-Hexol, 2,3,6,7,10,11-Triphenylenehexol

Chemical Identifiers

CAS 4877-80-9
Molecular Formula C18H12O6
Molecular Weight (g/mol) 324.29
InChI Key QMLILIIMKSKLES-UHFFFAOYSA-N
IUPAC Name triphenylene-2,3,6,7,10,11-hexol
SMILES OC1=C(O)C=C2C(=C1)C1=CC(O)=C(O)C=C1C1=CC(O)=C(O)C=C21

Specifications

Quantity 50 g
Percent Purity 95%
Physical Form Crystals or Flakes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.