accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 488-11-9

IUPAC Name: (2Z)-2,3-dibromo-4-oxobut-2-enoate
Synonyms: mucobromic acid
Molecular Weight (g/mol): 256.86
Molecular Formula: C4HBr2O3
InChi Key: NCNYEGJDGNOYJX-IHWYPQMZSA-M
SMILES: [O-]C(=O)C(\Br)=C(\Br)C=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)