Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mucobromic acid

Catalog No. M89625100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M89625100G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M89625100G Supplier Sigma Aldrich Supplier No. M89625100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-(trihydroxysilyl)propyl methylphosphonate, monosodium salt was used for the following syntheses:
  • Near-infrared dye-doped nanoparticles (nps)
  • As single silica source for the fabrication of phosphonate-rich organosilica layered hybrid material (pslm)
  • Gd/sio2 nanoparticles
  • Mesoporous silica nanoparticles
  • Cdse/zns/silica functionalized nanocrystals

Specifications

CAS 488-11-9
Melting Point 120°C to 124°C (lit.)
Molecular Formula C4H2Br2O3
Linear Formula OHCCBr=CBrCOOH
MDL Number MFCD00063745
Quantity 100 mL
Synonym 2,3-Dibromomalealdehydic acid
Molecular Weight (g/mol) 257.86
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.