accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 488-23-3

IUPAC Name: 1,2,3,4-tetramethylbenzene
Synonyms: 1,2,3,4-tetramethylbenzene prehnitene prehnitol
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: UOHMMEJUHBCKEE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(C)=C1C
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results