Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prehnitene, MP Biomedicals

Catalog No. ICN217115 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217115 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217115 Supplier MP Biomedicals, Inc Supplier No. 0521711505

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 488-23-3
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
InChI Key UOHMMEJUHBCKEE-UHFFFAOYSA-N
Synonym prehnitene, prehnitol, benzene, 1,2,3,4-tetramethyl, tetramethylbenzene, unii-96wt7d2wxj, benzene, tetramethyl, 96wt7d2wxj, ccris 8659, tetramethylbenzenes, tetra-methyl benzene
PubChem CID 10263
ChEBI CHEBI:38997
IUPAC Name 1,2,3,4-tetramethylbenzene
SMILES CC1=C(C(=C(C=C1)C)C)C

Specifications

Quantity 100 g
Formula Weight 134.2
Chemical Name or Material Prehnitene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.