accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 488-43-7

IUPAC Name: 6-aminohexane-1,2,3,4,5-pentol
Synonyms: glucamine
Molecular Weight (g/mol): 181.19
Molecular Formula: C6H15NO5
InChi Key: SDOFMBGMRVAJNF-UHFFFAOYNA-N
SMILES: NCC(O)C(O)C(O)C(O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results