accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)H3CCH3OO

CAS RN 4883-60-7

IUPAC Name: (3R)-3,5,5-trimethylcyclohexane-1,2-dione
Synonyms: (3R)-3,5,5-trimethylcyclohexane-1,2-dione
Molecular Weight (g/mol): 154.21
Molecular Formula: C9H14O2
InChi Key: WEUZZYGESXCTCU-ZCFIWIBFSA-N
SMILES: C[C@@H]1CC(C)(C)CC(=O)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results