Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-3,5,5-trimethyl-2-cyclohexen-1-one 98.0+%, TCI America™
SDP

Catalog No. T27235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27235G Supplier TCI America Supplier No. T27235G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4883-60-7
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD02181023
InChI Key DWGZTTFGUFHAJX-UHFFFAOYSA-N
Synonym 3,5,5-Trimethyl-1,2-cyclohexanedione, 2-Hydroxyisophorone
PubChem CID 551084
IUPAC Name 2-hydroxy-3,5,5-trimethylcyclohex-2-en-1-one
SMILES CC1=C(C(=O)CC(C1)(C)C)O

Specifications

Melting Point 92°C
Color White-Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 154.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-3,5,5-trimethyl-2-cyclohexen-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.