accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4885-02-3

IUPAC Name: dichloro(methoxy)methane
Synonyms: dichloromethyl methyl ether
Molecular Weight (g/mol): 114.95
Molecular Formula: C2H4Cl2O
InChi Key: GRTGGSXWHGKRSB-UHFFFAOYSA-N
SMILES: COC(Cl)Cl
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)