accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 489-01-0

IUPAC Name: 2,6-di-tert-butyl-4-methoxyphenol
Synonyms: 2,6-di-tert-butyl-4-methoxyphenol
Molecular Weight (g/mol): 236.36
Molecular Formula: C15H24O2
InChi Key: SLUKQUGVTITNSY-UHFFFAOYSA-N
SMILES: COC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)