accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNOHO

CAS RN 489446-85-7

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}cyclopentane-1-carboxylic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]cyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: RNJQBGXOSAQQDG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)