accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 490-83-5

IUPAC Name: 5-(1,2-dihydroxyethyl)oxolane-2,3,4-trione
Synonyms: dehydroascorbic acid 1-dehydroascorbate L-ascorbic acid, dehydro- acid, dehydroascorbic oxidized ascorbate dehydroascorbates 1-dehydroascorbic acid
Molecular Weight (g/mol): 174.11
Molecular Formula: C6H6O6
InChi Key: SBJKKFFYIZUCET-UHFFFAOYNA-N
SMILES: OCC(O)C1OC(=O)C(=O)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)