CAS RN 490-91-5
Thymoquinone, 99%
Thymoquinone
CAS: 490-91-5 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.20 InChI Key: KEQHJBNSCLWCAE-UHFFFAOYSA-N IUPAC Name: 2-methyl-5-(propan-2-yl)cyclohexa-2,5-diene-1,4-dione SMILES: CC(C)C1=CC(=O)C(C)=CC1=O
Thymoquinone 98.0+%, TCI America™
CAS: 490-91-5 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 MDL Number: MFCD00001602 InChI Key: KEQHJBNSCLWCAE-UHFFFAOYSA-N Synonym: thymoquinone,thymoquinon,p-cymene-2,5-dione,2-isopropyl-5-methylbenzoquinone,polythymoquinone,2,5-cyclohexadiene-1,4-dione, 2-methyl-5-1-methylethyl,2-isopropyl-5-methyl-1,4-benzoquinone,5-isopropyl-2-methyl-1,4-benzoquinone,2-isopropyl-5-methyl-p-benzoquinone,p-mentha-3,6-diene-2,5-dione PubChem CID: 10281 IUPAC Name: 2-methyl-5-propan-2-ylcyclohexa-2,5-diene-1,4-dione SMILES: CC1=CC(=O)C(=CC1=O)C(C)C
MP Biomedicals, Inc Thymoquinone, MP Biomedicals™
CAS: 490-91-5 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 InChI Key: KEQHJBNSCLWCAE-UHFFFAOYSA-N Synonym: thymoquinone,thymoquinon,p-cymene-2,5-dione,2-isopropyl-5-methylbenzoquinone,polythymoquinone,2,5-cyclohexadiene-1,4-dione, 2-methyl-5-1-methylethyl,2-isopropyl-5-methyl-1,4-benzoquinone,5-isopropyl-2-methyl-1,4-benzoquinone,2-isopropyl-5-methyl-p-benzoquinone,p-mentha-3,6-diene-2,5-dione PubChem CID: 10281 IUPAC Name: 2-methyl-5-propan-2-ylcyclohexa-2,5-diene-1,4-dione SMILES: CC1=CC(=O)C(=CC1=O)C(C)C