Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymoquinone, MP Biomedicals™

Catalog No. ICN15691801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15691801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15691801 Supplier MP Biomedicals, Inc Supplier No. 0215691801

Chemical Identifiers

CAS 490-91-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
InChI Key KEQHJBNSCLWCAE-UHFFFAOYSA-N
Synonym thymoquinone, thymoquinon, p-cymene-2,5-dione, 2-isopropyl-5-methylbenzoquinone, polythymoquinone, 2,5-cyclohexadiene-1,4-dione, 2-methyl-5-1-methylethyl, 2-isopropyl-5-methyl-1,4-benzoquinone, 5-isopropyl-2-methyl-1,4-benzoquinone, 2-isopropyl-5-methyl-p-benzoquinone, p-mentha-3,6-diene-2,5-dione
PubChem CID 10281
IUPAC Name 2-methyl-5-propan-2-ylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C(=CC1=O)C(C)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 164.2
Physical Form Crystalline Powder
Chemical Name or Material Thymoquinone, For TLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.