CAS RN 4911-54-0
IUPAC Name:
ethyl 4-methylpent-4-enoate
Synonyms:
ethyl 4-methylpent-4-enoate
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
RRTBSPBUHUUTHR-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCC(C)=C
1
–
1
of
1
results
1
Sigma Aldrich Ethyl 4-methyl-4-pentenoate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.