accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3H3C

CAS RN 4913-13-7

IUPAC Name: N,N,3,5-tetramethylaniline
Synonyms: N,N,3,5-tetramethylaniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: NBFRQCOZERNGEX-UHFFFAOYSA-N
SMILES: CN(C)C1=CC(C)=CC(C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)