Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,3,5-Tetramethylaniline 98.0+%, TCI America™
SDP

Catalog No. T24725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24725G Supplier TCI America Supplier No. T24725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4913-13-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008307
InChI Key NBFRQCOZERNGEX-UHFFFAOYSA-N
Synonym benzenamine, n,n,3,5-tetramethyl, n,n-dimethyl-3,5-xylidine, 3,5-xylidine, n,n-dimethyl, 3,5-dimethylphenyl dimethylamine, acmc-209kew, 5-dimethylamino-m-xylene, 3,5,n,n-tetramethylaniline, n,n-dimethyl-sym-m-xylidine, n,n,3,5-tetramethyl aniline, n,n-3,5-tetramethylbenzenamine
PubChem CID 78625
IUPAC Name N,N,3,5-tetramethylaniline
SMILES CC1=CC(=CC(=C1)N(C)C)C

Specifications

Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2810
Boiling Point 224°C
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material N,N,3,5-Tetramethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.