accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4914-91-4

IUPAC Name: (2Z)-3,4-dimethylpent-2-ene
Synonyms: 2-pentene, 3,4-dimethyl-, (Z)-
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: PPBWEVVDSRKEIK-ALCCZGGFSA-N
SMILES: C\C=C(\C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results