Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3,4-Dimethyl-2-pentene 98.0+%, TCI America™
SDP

Catalog No. D12555ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12555ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12555ML Supplier TCI America Supplier No. D12555ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4914-91-4
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00059237
InChI Key PPBWEVVDSRKEIK-ALCCZGGFSA-N
PubChem CID 643784
IUPAC Name (Z)-3,4-dimethylpent-2-ene
SMILES CC=C(C)C(C)C

Specifications

Color Colorless
Boiling Point 87°C
Quantity 5 mL
UN Number 3295
Formula Weight 98.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3,4-Dimethyl-2-pentene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.