accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHO

CAS RN 492-88-6

IUPAC Name: 3-ethoxy-2-hydroxybenzaldehyde
Synonyms: 3-ethoxy-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: OFQBYHLLIJGMNP-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(C=O)=C1O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)