Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxysalicylaldehyde 97.0+%, TCI America™
SDP

Catalog No. E025325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E025325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E025325G Supplier TCI America Supplier No. E025325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 492-88-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003323
InChI Key OFQBYHLLIJGMNP-UHFFFAOYSA-N
Synonym 3-ethoxysalicylaldehyde, benzaldehyde, 3-ethoxy-2-hydroxy, novovanillin, o-ethylvanillin, salicylaldehyde, 3-ethoxy, unii-hb4h48mkni, 2-hydroxy-3-ethoxybenzaldehyde, hb4h48mkni, 3-ethoxy-2-hydroxy-benzaldehyde, pubchem3123
PubChem CID 68117
IUPAC Name 3-ethoxy-2-hydroxybenzaldehyde
SMILES CCOC1=CC=CC(=C1O)C=O

Specifications

Melting Point 66°C
Boiling Point 264°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethoxysalicylaldehyde
RTECSNumber CU6123000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.