accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONHNOO

CAS RN 4926-55-0

IUPAC Name: 2-[(2-nitrophenyl)amino]ethan-1-ol
Synonyms: HC yellow NO 2
Molecular Weight (g/mol): 182.18
Molecular Formula: C8H10N2O3
InChi Key: LFOUYKNCQNVIGI-UHFFFAOYSA-N
SMILES: OCCNC1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)