Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Nitroanilino)ethanol 98.0+%, TCI America™
SDP

Catalog No. N10005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10005G 5 g
N100025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10005G Supplier TCI America Supplier No. N10005G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4926-55-0
Molecular Formula C8H10N2O3
Molecular Weight (g/mol) 182.179
MDL Number MFCD00239475
InChI Key LFOUYKNCQNVIGI-UHFFFAOYSA-N
Synonym N-(2-Hydroxyethyl)-2-nitroaniline, N-(2-Nitrophenyl)ethanolamine
PubChem CID 78637
IUPAC Name 2-(2-nitroanilino)ethanol
SMILES C1=CC=C(C(=C1)NCCO)[N+](=O)[O-]

Specifications

Melting Point 73°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Nitroanilino)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.