CAS RN 493028-20-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ CBFβ Inhibitor
The CBFβ Inhibitor, also referenced under CAS 493028-20-9, controls the biological activity of CBFβ.