accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4931-66-2

IUPAC Name: N'-methyl-N-[3-(trifluoromethyl)phenyl]{[(4-isothiocyanatophenyl)methyl]sulfanyl}methanimidamide
Synonyms: N'-methyl-N-[3-(trifluoromethyl)phenyl][(4-isothiocyanatophenyl)methyl]sulfanylmethanimidamide
Molecular Weight (g/mol): 381.44
Molecular Formula: C17H14F3N3S2
InChi Key: JLGLYOGXDCENSE-UHFFFAOYSA-N
SMILES: CN=C(NC1=CC=CC(=C1)C(F)(F)F)SCC1=CC=C(C=C1)N=C=S
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)