Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate, 97%

Catalog No. AAH6254322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6254322 100 g
AAH6254306 5 g
AAH6254314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6254322 Supplier Thermo Scientific Chemicals Supplier No. H6254322
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4931-66-2 | C17H14F3N3S2 | 381.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4931-66-2
Molecular Formula C17H14F3N3S2
Molecular Weight (g/mol) 381.44
MDL Number MFCD00080931
InChI Key JLGLYOGXDCENSE-UHFFFAOYSA-N
Synonym methyl l-pyroglutamate, methyl s-+-2-pyrrolidone-5-carboxylate, l-pyroglutamic acid methyl ester, methyl pyroglutamate, methyl 5-oxo-l-prolinate, s-methyl 5-oxopyrrolidine-2-carboxylate, proline, 5-oxo-, methyl ester, methyl 2s-5-oxopyrrolidine-2-carboxylate, h-pyr-ome, s-+-methyl 2-pyrrolidone-5-carboxylate
PubChem CID 78646
IUPAC Name methyl (2S)-5-oxopyrrolidine-2-carboxylate
SMILES CN=C(NC1=CC=CC(=C1)C(F)(F)F)SCC1=CC=C(C=C1)N=C=S

Specifications

Color Yellow
Density 1.226 g/mL
Boiling Point 90°C (0.3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.486
Quantity 100 g
Formula Weight 143.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-567-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.