accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH

CAS RN 494-19-9

IUPAC Name: 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
Synonyms: 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
Molecular Weight (g/mol): 195.27
Molecular Formula: C14H13N
InChi Key: ZSMRRZONCYIFNB-UHFFFAOYSA-N
SMILES: C1CC2=CC=CC=C2NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)