CAS RN 494227-37-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sigma Aldrich (Sp,S'P)-1,1'-Bis[Bis(4-Methoxy-3,5-Dimethylphenyl)Phosphino]-2,2'-Bis[™-Alpha-(Dimethylamino)Benzyl]Ferrocene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.