Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Sp,S'P)-1,1'-Bis[Bis(4-Methoxy-3,5-Dimethylphenyl)Phosphino]-2,2'-Bis[™-Alpha-(Dimethylamino)Benzyl]Ferrocene

Catalog No. 7346950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-469-50G 50 g
73-469-100G 100 g
73-469-1G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 73-469-50G Supplier Sigma Aldrich Supplier No. 7346950G
Only null left
Add to Cart
Add to Cart
Request a Quote

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mandyphos SL-M004-1 is a chiral ferrocenyl based diphosphine ligand belonging to the Mandyphos family.It may be used as a chiral ligand in the rhodium complex-catalyzed hydrogenation reaction, a key step during the synthesis of argatroban.sold in collaboration with Solvias AG

Specifications

CAS 494227-37-1
Molecular Formula C64H74FeN2O4P2
Linear Formula C64H74FeN2O4P2
MDL Number MFCD04117704
Quantity 50 g
Synonym (1R,1'R)- 1,1'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2'-bis[(R)-(dimethylamino)phenylmethyl]ferrocene (acc to CAS); Mandyphos SL-M004-1
Molecular Weight (g/mol) 1053.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.