accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS

CAS RN 4946-13-8

IUPAC Name: (4-ethylphenyl)sulfanide
Synonyms: (4-ethylphenyl)sulfanide
Molecular Weight (g/mol): 137.22
Molecular Formula: C8H9S
InChi Key: WWQQPHUHTAZWDH-UHFFFAOYSA-M
SMILES: CCC1=CC=C([S-])C=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)