Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylbenzenethiol, TCI America™
SDP

Catalog No. E067225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E067225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E067225G Supplier TCI America Supplier No. E067225G

missing translation for 'validMainframeMsg'

Specifications

CAS 4946-13-8
Color Yellow
Boiling Point 211°C
Molecular Formula C8H9S
MDL Number MFCD00040229
Quantity 25 g
Synonym 4-ethylthiophenol, 4-ethyl thiophenol, 4-ethylbenzene-1-thiol, benzenethiol, 4-ethyl, p-ethylphenylmercaptan, p-ethylthiophenol, pubchem6813, p-ethylphenyl mercaptan, acmc-209kg5
InChI Key WWQQPHUHTAZWDH-UHFFFAOYSA-M
SMILES CCC1=CC=C([S-])C=C1
IUPAC Name (4-ethylphenyl)sulfanide
Molecular Weight (g/mol) 137.22
PubChem CID 2758914
Formula Weight 138.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylbenzenethiol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.