CAS RN 494772-07-5
CAS RN 494772-07-5
IUPAC Name:
5-bromo-2-(oxan-4-yloxy)pyridine
Synonyms:
5-bromo-2-(oxan-4-yloxy)pyridine
Molecular Weight (g/mol):
258.12
Molecular Formula:
C10H12BrNO2
InChi Key:
ZCTXTBRJCPHMEP-UHFFFAOYSA-N
SMILES:
BrC1=CN=C(OC2CCOCC2)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 5-Bromo-2-(tetrahydropyran-4-yloxy)pyridine, 97%, Thermo Scientific™
CAS: 494772-07-5 Molecular Formula: C10H12BrNO2 Molecular Weight (g/mol): 258.115 MDL Number: MFCD09064985 InChI Key: ZCTXTBRJCPHMEP-UHFFFAOYSA-N Synonym: 5-bromo-2-tetrahydro-2h-pyran-4-yloxy pyridine,5-bromo-2-tetrahydro-2h-pyran-4-yl oxy pyridine,5-bromo-2-tetrahydropyran-4-yloxy pyridine,5-bromo-2-oxan-4-yloxy pyridine,pyridine,5-bromo-2-tetrahydro-2h-pyran-4-yl oxy,pubchem22061,5-bromo-2-oxan-4-yl oxy pyridine,2-tetrahydropyran-4-yloxy-5-bromopridine,2-tetrahydropyran-4-yloxy-5-bromopyridine,4-5-bromopyridin-2-yl oxy oxane PubChem CID: 21914391 IUPAC Name: 5-bromo-2-(oxan-4-yloxy)pyridine SMILES: C1COCCC1OC2=NC=C(C=C2)Br