accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 495-78-3

IUPAC Name: 3-(2-hydroxyphenyl)propanoic acid
Synonyms: melilotic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: CJBDUOMQLFKVQC-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results