Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β(o-Hydroxyphenyl)propionic Acid,MP Biomedicals

Catalog No. ICN227208 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227208 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227208 Supplier MP Biomedicals, Inc Supplier No. 0522720810

Chemical Identifiers

CAS 495-78-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
InChI Key CJBDUOMQLFKVQC-UHFFFAOYSA-N
Synonym 3-2-hydroxyphenyl propanoic acid, 3-2-hydroxyphenyl propionic acid, melilotic acid, benzenepropanoic acid, 2-hydroxy, melilotate, o-hydroxyphenylpropionic acid, 2-hydroxybenzenepropanoic acid, 3-o-hydroxyphenyl propionic acid, 2-hydroxyphenylpropanoate, unii-cn1rk42mad
PubChem CID 873
ChEBI CHEBI:16104
IUPAC Name 3-(2-hydroxyphenyl)propanoic acid
SMILES C1=CC=C(C(=C1)CCC(=O)O)O

Specifications

Quantity 10 g
Formula Weight 166.18
Chemical Name or Material Beta-(O-Hydroxyphenyl)Propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.