accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOH3CCH3OOCl

CAS RN 49562-28-9

IUPAC Name: propan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
Synonyms: fenofibrate ankebin fenobrate fenofibrate preparation fournier brand of procetofen lipidil ter hexal brand of procetofen novopharm brand of procetofen lipofen lipofene
Molecular Weight (g/mol): 360.83
Molecular Formula: C20H21ClO4
InChi Key: YMTINGFKWWXKFG-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C(C)(C)OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (1)
Grade 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)