Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenofibrate, BP, EP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602655 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-655 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-602-655 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F360225KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

F3602, 49562-28-9, C20H21ClO4

Spectrum™ Chemical Fenofibrate, BP, EP

Chemical Identifiers

CAS 49562-28-9
Molecular Formula C20H21ClO4
Molecular Weight (g/mol) 360.83
InChI Key YMTINGFKWWXKFG-UHFFFAOYSA-N
IUPAC Name propan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 98 to 102%
Grade Multi Compendia
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.