CAS RN 49573-30-0
CAS RN 49573-30-0
IUPAC Name:
quinoline-7-carbaldehyde
Synonyms:
quinoline-7-carbaldehyde
Molecular Weight (g/mol):
157.17
Molecular Formula:
C10H7NO
InChi Key:
WINWAFCAQPFBQA-UHFFFAOYSA-N
SMILES:
O=CC1=CC2=NC=CC=C2C=C1
1
–
1
of
1
results
1
7-Quinolinecarboxaldehyde 98.0+%, TCI America™
CAS: 49573-30-0 Molecular Formula: C10H7NO Molecular Weight (g/mol): 157.172 MDL Number: MFCD01366121 InChI Key: WINWAFCAQPFBQA-UHFFFAOYSA-N Synonym: 7-Formylquinoline PubChem CID: 4712255 IUPAC Name: quinoline-7-carbaldehyde SMILES: C1=CC2=C(C=C(C=C2)C=O)N=C1