Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Quinolinecarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. Q0067250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0067250MG 250 mg
Q00671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q0067250MG Supplier TCI America Supplier No. Q0067250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 49573-30-0
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD01366121
InChI Key WINWAFCAQPFBQA-UHFFFAOYSA-N
Synonym 7-Formylquinoline
PubChem CID 4712255
IUPAC Name quinoline-7-carbaldehyde
SMILES C1=CC2=C(C=C(C=C2)C=O)N=C1

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 250 mg
Formula Weight 157.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Quinolinecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.