accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49590-51-4

IUPAC Name: 2-(2-formylphenoxy)benzaldehyde
Synonyms: 2-(2-formylphenoxy)benzaldehyde
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: LMJZLKFWMQOYKU-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1OC1=CC=CC=C1C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results