Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-formylphenyl) Ether 98.0+%, TCI America™
SDP

Catalog No. B16461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16461G 1 g
B164610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16461G Supplier TCI America Supplier No. B16461G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 49590-51-4
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00191616
InChI Key LMJZLKFWMQOYKU-UHFFFAOYSA-N
PubChem CID 614688
IUPAC Name 2-(2-formylphenoxy)benzaldehyde
SMILES C1=CC=C(C(=C1)C=O)OC2=CC=CC=C2C=O

Specifications

Melting Point 78°C
Color White-Yellow
Boiling Point 182°C
Quantity 1 g
Synonym 2,2′C-Diformyldiphenyl Ether, 2,2′C-Oxydibenzaldehyde
Formula Weight 226.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-formylphenyl) Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.