accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 496-16-2

IUPAC Name: 2,3-dihydro-1-benzofuran
Synonyms: 2,3-dihydrobenzofuran
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: HBEDSQVIWPRPAY-UHFFFAOYSA-N
SMILES: C1CC2=CC=CC=C2O1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)