CAS RN 496864-16-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Aloisine A
A cell-permeable pyrrolo-pyrazine compound that exerts anti-proliferative effects.