accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHH3CCH3H3CCH3H3CCH3

CAS RN 497-39-2

IUPAC Name: 2,4-di-tert-butyl-5-methylphenol
Synonyms: 2,4-di-tert-butyl-5-methylphenol
Molecular Weight (g/mol): 220.36
Molecular Formula: C15H24O
InChi Key: WYSSJDOPILWQDC-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(C=C1C(C)(C)C)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results