accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4971-55-5

IUPAC Name: 3-chlorooxolane-2,4-dione
Synonyms: 3-chlorooxolane-2,4-dione
Molecular Weight (g/mol): 134.52
Molecular Formula: C4H3ClO3
InChi Key: DJEJYZZJYYNADA-UHFFFAOYNA-N
SMILES: ClC1C(=O)COC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)