Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,4(3H,5H)-furandione 98.0+%, TCI America™
SDP

Catalog No. C18105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18105G 5 g
C181025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18105G Supplier TCI America Supplier No. C18105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4971-55-5
Molecular Formula C4H3ClO3
Molecular Weight (g/mol) 134.52
MDL Number MFCD00134273
InChI Key DJEJYZZJYYNADA-UHFFFAOYNA-N
Synonym 3-chloro-2,4 3h,5h-furandione, 3-chlorofuran-2,4 3h,5h-dione, 3-chloro-furan-2,4-dione, 3-chlorotetronic acid, 2,4 3h,5h-furandione, 3-chloro, 3-chloro-3,5-dihydrofuran-2,4-dione, acmc-209khp, 3-chlorofuran-2,4-dione, 3-chloranyloxolane-2,4-dione, 3-chlorotetrahydrofuran-2,4-dione
PubChem CID 4158575
IUPAC Name 3-chlorooxolane-2,4-dione
SMILES ClC1C(=O)COC1=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 134.52
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2,4(3H,5H)-furandione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.