accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 497147-93-0

IUPAC Name: (5-cyanopyridin-3-yl)boronic acid
Synonyms: 5-cyanopyridin-3-ylboronic acid
Molecular Weight (g/mol): 147.93
Molecular Formula: C6H5BN2O2
InChi Key: CYEXXDYQJPRMIQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CN=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results