accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3NH2NNH3H3NH2NNH3OHPOOOOHPOOOOHPOOO

CAS RN 49721-50-8

IUPAC Name: bis((4-azaniumylbutyl)(3-azaniumylpropyl)azanium) trihydrogen phosphate
Synonyms: bis(spermidine(3+)) trihydrogen phosphate
Molecular Weight (g/mol): 584.48
Molecular Formula: C14H47N6O12P3
InChi Key: RQFVSMCFDFGRDX-UHFFFAOYSA-N
SMILES: OP([O-])([O-])=O.OP([O-])([O-])=O.OP([O-])([O-])=O.[NH3+]CCCC[NH2+]CCC[NH3+].[NH3+]CCCC[NH2+]CCC[NH3+]
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results