Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermidine Phosphate Hexahydrate 98.0+%, TCI America™
SDP

Catalog No. S03851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03851G Supplier TCI America Supplier No. S03851G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49721-50-8
Molecular Formula C14H38N6
MDL Number MFCD00043283
Synonym N-(3-Aminopropyl)-1,4-butanediamine Phosphate, N-(3-Aminopropyl)tetramethylenediamine Phosphate

Specifications

Color White
Quantity 1 g
Formula Weight 584.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Spermidine Phosphate Hexahydrate
RTECSNumber EJ7020000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.