accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49732-38-9

IUPAC Name: 4-[(4-methoxyphenyl)amino]benzene-1-diazonium hydrogen sulfate
Synonyms: 4-[(4-methoxyphenyl)amino]benzenediazonium hydrogen sulfate
Molecular Weight (g/mol): 323.32
Molecular Formula: C13H13N3O5S
InChi Key: UXVMTJSLLAUFTO-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.COC1=CC=C(NC2=CC=C(C=C2)[N+]#N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results